missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butylurea, 96%
Description
n-Butylurea is used as a fertilizer for rice. It is used in the preparation of drugs which are recommended for diabetes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Butylurea is used as a fertilizer for rice. It is used in the preparation of drugs which are recommended for diabetes.
Solubility
Soluble in water, Alcohol, Ether.
Notes
Incompatible with Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
n-Butylurea is used as a fertilizer for rice. It is used in the preparation of drugs which are recommended for diabetes.
Solubility
Soluble in water, Alcohol, Ether.
Notes
Incompatible with Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
Specifications
Specifications
| Melting Point | 92°C to 96°C |
| Quantity | 100 g |
| Beilstein | 1744775 |
| Solubility Information | Soluble in water,Alcohol,Ether. |
| Formula Weight | 116.16 |
| Percent Purity | 96% |
| Chemical Name or Material | n-Butylurea |
Safety and Handling
| Hazard Category | H302+H312+H332-H335-H341 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c |
| EINECSNumber | 209-748-4 |
| RTECSNumber | YS3675000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only