missing translation for 'onlineSavingsMsg'
Learn More

n-Butylurea, 96%

Catalog No. AAL0285422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0285422 100 g
AAL0285436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0285422 Supplier Thermo Scientific Chemicals Supplier No. L0285422

CAS: 592-31-4 | C5H12N2O | 116.164 g/mol

n-Butylurea is used as a fertilizer for rice. It is used in the preparation of drugs which are recommended for diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butylurea is used as a fertilizer for rice. It is used in the preparation of drugs which are recommended for diabetes.

Solubility
Soluble in water, Alcohol, Ether.

Notes
Incompatible with Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.

Chemical Identifiers

CAS 592-31-4
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.164
MDL Number MFCD00007956
InChI Key CNWSQCLBDWYLAN-UHFFFAOYSA-N
Synonym n-butylurea, 1-butylurea, urea, butyl, urea, n-butyl, unii-9cpl5nr15k, n-butyl urea, 9cpl5nr15k, dsstox_cid_222, dsstox_rid_75443, dsstox_gsid_20222
PubChem CID 11595
IUPAC Name butylurea
SMILES CCCCNC(=O)N

Specifications

Melting Point 92°C to 96°C
Quantity 100 g
Beilstein 1744775
Solubility Information Soluble in water,Alcohol,Ether.
Formula Weight 116.16
Percent Purity 96%
Chemical Name or Material n-Butylurea
Hazard Category H302+H312+H332-H335-H341
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
EINECSNumber 209-748-4
RTECSNumber YS3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only