Learn More
n-Butyltin hydroxide oxide, 95%
CAS: 2273-43-0 | C4H10O2Sn | 208.83 g/mol
Supplier: Thermo Scientific Chemicals A1829036
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| n-Butyltin hydroxide oxide | |
| C4H10O2Sn | |
| 500 g | |
| Hygroscopic | |
| WIHMDCQAEONXND-UHFFFAOYSA-M | |
| butyl-hydroxy-oxotin | |
| 16767 | |
| 95% |
| 2273-43-0 | |
| MFCD00013927 | |
| UN3146 | |
| stannane, butylhydroxyoxo, butyltin hydroxide oxide, butylstannoic acid, butylstannonic acid, 1-butanestannonic acid, n-butyltin hydroxide oxide, butylhydroxytin oxide, butylhydroxyoxostannane, unii-g34wda7z2e, butylenestannonic acid | |
| CCCC[Sn](O)=O | |
| 208.83 | |
| 208.81 |
Chemical Identifiers
| 2273-43-0 | |
| 208.83 | |
| WIHMDCQAEONXND-UHFFFAOYSA-M | |
| 16767 |
| C4H10O2Sn | |
| MFCD00013927 | |
| stannane, butylhydroxyoxo, butyltin hydroxide oxide, butylstannoic acid, butylstannonic acid, 1-butanestannonic acid, n-butyltin hydroxide oxide, butylhydroxytin oxide, butylhydroxyoxostannane, unii-g34wda7z2e, butylenestannonic acid | |
| CCCC[Sn](O)=O |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber : 218-880-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only