missing translation for 'onlineSavingsMsg'
Learn More

n-Butyltin hydroxide oxide, 95%

Catalog No. AAA1829022
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1829022 100 g
AAA1829036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1829022 Supplier Thermo Scientific Chemicals Supplier No. A1829022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2273-43-0 | C4H10O2Sn | 208.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2273-43-0
Molecular Formula C4H10O2Sn
Molecular Weight (g/mol) 208.83
MDL Number MFCD00013927
InChI Key WIHMDCQAEONXND-UHFFFAOYSA-M
Synonym stannane, butylhydroxyoxo, butyltin hydroxide oxide, butylstannoic acid, butylstannonic acid, 1-butanestannonic acid, n-butyltin hydroxide oxide, butylhydroxytin oxide, butylhydroxyoxostannane, unii-g34wda7z2e, butylenestannonic acid
PubChem CID 16767
SMILES CCCC[Sn](O)=O

Specifications

Quantity 100 g
UN Number UN3146
Sensitivity Hygroscopic
IUPAC Name butyl(hydroxy)stannanone
Formula Weight 208.81
Percent Purity 95%
Chemical Name or Material n-Butyltin hydroxide oxide
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 218-880-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only