missing translation for 'onlineSavingsMsg'
Learn More

N-Butylphthalimide 98.0+%, TCI America™

Catalog No. B073625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B073625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B073625G Supplier TCI America Supplier No. B073625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1515-72-6
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00039695
InChI Key DLKDEVCJRCPTLN-UHFFFAOYSA-N
Synonym n-butylphthalimide, 2-butylisoindoline-1,3-dione, n-n-butyl phthalimide, 1-phthalimidobutane, phthalimide, n-butyl, 1h-isoindole-1,3 2h-dione, 2-butyl, unii-5th1dkt35e, 2-butyl-1h-isoindole-1,3 2h-dione, 5th1dkt35e, 2-butylbenzo c azoline-1,3-dione
PubChem CID 73812
IUPAC Name 2-butylisoindole-1,3-dione
SMILES CCCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 203.24
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material N-Butylphthalimide