missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich N-Butyllithium Solution

Catalog No. 503081439
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
503081439 100 mL
186171800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 503081439 Supplier Sigma Aldrich Supplier No. 186171100ML
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

May contain trace amounts of LiCl.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 109-72-8
Density 0.68 g/mL at 25°C
Molecular Formula C4H9Li
Linear Formula CH3(CH2)3Li
MDL Number MFCD00009414
Quantity 100 mL
Synonym n-BuLi; Butyl lithium; Butyllithium solution; Lithium-1-butanide
Molecular Weight (g/mol) 64.06 g/mol
Biological Activity Central Nervous System