missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich N-Butyllithium Solution

Catalog No. 503081483
Encompass
Change view
Click to view available options
Quantity:
800 mL
4 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
503081483 4 x 100 mL
230707800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 503081483 Supplier Sigma Aldrich Supplier No. 2307074X100ML
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 109-72-8
Density 0.693 g/mL at 25°C
Molecular Formula C4H9Li
Linear Formula CH3(CH2)3Li
MDL Number MFCD00009414
Quantity 4 x 100 mL
Synonym n-BuLi; Butyl lithium; Butyllithium solution; Lithium-1-butanide
Molecular Weight (g/mol) 64.06 g/mol
Biological Activity Central Nervous System