missing translation for 'onlineSavingsMsg'
Learn More

n-Butyllithium, 2.5M solution in hexanes, AcroSeal™

Catalog No. AC213350500
Change view
Click to view available options
Quantity:
50 mL
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC213350500 50 mL Glass bottle
AC213351000 100 mL AcroSeal™ Glass bottle
AC213358000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC213350500 Supplier Thermo Scientific Chemicals Supplier No. 213350500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-72-8 | C4H9Li | 64.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 109-72-8
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00009414
InChI Key MZRVEZGGRBJDDB-UHFFFAOYSA-N
Synonym unii-09w9a6b8zc, buli, lithium, butyl, n-buli, n-butyllithium, n-butyl lithium, butyllithium, lithium butane, libu, butyl lithium
PubChem CID 61028
SMILES [Li]CCCC

Specifications

Melting Point -95.0°C
Density 0.6900 g/mL
Boiling Point 60.0°C to 80.0°C
Flash Point -21°C
Packaging Glass bottle
Linear Formula CH3(CH2)3Li
Quantity 50 mL
Fieser 01,95; 02,51; 04,60; 05,78; 06,85; 07,45; 08,65; 09,83; 10,68; 11,101; 12,96; 13,56; 14,63; 15,59; 16,55; 17,59
Specific Gravity 0.69
IUPAC Name lithium(1+) butan-1-ide
Formula Weight 64.06
Chemical Name or Material n-Butyllithium, 2.5 M Solution in Hexanes
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. May cause drowsiness or dizziness. Suspected of damaging fertility. Highly flammable liquid and vapor. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Store in a dry place. Store in a closed container. Ground/Bond container and receiving equipment. Take precautionary measures against static discharge.
EINECSNumber 203-698-7
TSCA TSCA

RUO – Research Use Only