missing translation for 'onlineSavingsMsg'
Learn More

N-Butylimidazole, 98%

CAS: 4316-42-1 | C7H12N2 | 124.19 g/mol

$522.01 - $522.01

Chemical Identifiers

CAS 4316-42-1
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00042753
InChI Key MCMFEZDRQOJKMN-UHFFFAOYSA-N
Synonym 1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
PubChem CID 61347
IUPAC Name 1-butyl-1H-imidazole
SMILES CCCCN1C=CN=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442771000
SDS View Documents
Thermo Scientific Chemicals
442771000
100 g
Each for $522.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4316-42-1
124.19
MCMFEZDRQOJKMN-UHFFFAOYSA-N
61347
CCCCN1C=CN=C1
C7H12N2
MFCD00042753
1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
1-butyl-1H-imidazole

Specifications

4316-42-1
257.0°C
Authentic
MFCD00042753
1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
MCMFEZDRQOJKMN-UHFFFAOYSA-N
1-butyl-1H-imidazole
61347
98%
0.9450g/mL
113°C
C7H12N2
100 g
Solubility in water: soluble.
CCCCN1C=CN=C1
124.19
124.19
N-Butylimidazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Do not breathe dust/fume/gas/mist/vapors/spray.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only