missing translation for 'onlineSavingsMsg'
Learn More

N-Butylimidazole, 98%

Catalog No. AC442770250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442770250 25 g
AC442771000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442770250 Supplier Thermo Scientific Chemicals Supplier No. 442770250

CAS: 4316-42-1 | C7H12N2 | 124.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4316-42-1
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00042753
InChI Key MCMFEZDRQOJKMN-UHFFFAOYSA-N
Synonym 1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
PubChem CID 61347
IUPAC Name 1-butyl-1H-imidazole
SMILES CCCCN1C=CN=C1

Specifications

Density 0.9450g/mL
Boiling Point 257.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Specific Gravity 0.945
Solubility Information Solubility in water: soluble.
Formula Weight 124.19
Percent Purity 98%
Chemical Name or Material N-Butylimidazole
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat
EINECSNumber 224-335-9

RUO – Research Use Only