missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyldimethylchlorosilane, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 138°C |
| Quantity | 100 g |
| Flash Point | 27°C (80°F) |
| Refractive Index | 1.42 |
| Sensitivity | Moisture sensitive |
| Density | 0.875 |
| UN Number | UN2920 |
| Beilstein | 1732573 |
| IUPAC Name | butyl-chloro-dimethylsilane |
| Formula Weight | 150.73 |
| Show More |
Safety and Handling
| Hazard Category | H226-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H226 Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S. |
| EINECSNumber | 423-700-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only