missing translation for 'onlineSavingsMsg'
Learn More

N-Butyldimethylamine 98.0+%, TCI America™

Catalog No. D150625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D150625ML 25 mL
D1506500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D150625ML Supplier TCI America Supplier No. D150625ML
Only null left

Chemical Identifiers

CAS 927-62-8
Molecular Formula C6H16N
Molecular Weight (g/mol) 102.20
MDL Number MFCD00043838
InChI Key DJEQZVQFEPKLOY-UHFFFAOYSA-O
Synonym N,N-Dimethylbutylamine, (Dimethylamino)butane
PubChem CID 13563
ChEBI CHEBI:59023
IUPAC Name butyldimethylazanium
SMILES CCCC[NH+](C)C

Specifications

Color Colorless
Boiling Point 93°C
Quantity 25 mL
UN Number 2733
Formula Weight 101.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Butyldimethylamine
EINECSNumber (2)-3356
RTECSNumber EJ4039250
TSCA Yes