missing translation for 'onlineSavingsMsg'
Learn More

N-Butyldiethanolamine 98.0+%, TCI America™

Catalog No. p-7118640
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B072525ML 25 mL
B0725500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B072525ML Supplier TCI America Supplier No. B072525ML
Only null left

Chemical Identifiers

CAS 102-79-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.245
MDL Number MFCD00002856
InChI Key GVNHOISKXMSMPX-UHFFFAOYSA-N
Synonym n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
PubChem CID 7620
IUPAC Name 2-[butyl(2-hydroxyethyl)amino]ethanol
SMILES CCCCN(CCO)CCO

Specifications

Melting Point -70°C
Color Yellow
Boiling Point 275°C
Quantity 25 mL
UN Number 2735
Formula Weight 161.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Butyldiethanolamine
EINECSNumber (2)-0334
RTECSNumber KK0525000
TSCA Yes