missing translation for 'onlineSavingsMsg'
Learn More

N-Butyldeoxynojirimycin, 98%, Thermo Scientific Chemicals

An →→alpha-glucosidase Inhibitor

Supplier:  Thermo Scientific Chemicals J66613MCR

Quantity 1 mg

 View more versions of this product

Catalog No. AAJ66613MCR


Only null left

Description

Description

An --alpha-glucosidase Inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

72599-27-0
219.28
UQRORFVVSGFNRO-UTINFBMNSA-N
CHEBI:50381
CCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO
C10H21NO4
MFCD00272581
51634
(2R,3R,4R,5S)-1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol

Specifications

N-Butyldeoxynojirimycin
Powder
126°C to 127°C
1 mg
MFCD00272581
Soluble in water at 10mg/mL
CCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO
219.28
CHEBI:50381
98%
N-Butyldeoxynojirimycin
72599-27-0
White
C10H21NO4
14,6188
UQRORFVVSGFNRO-UTINFBMNSA-N
(2R,3R,4R,5S)-1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol
51634
219.28
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TN4350150

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions