missing translation for 'onlineSavingsMsg'
Learn More

n-Butylcyclohexane, 99%

Catalog No. AAL0884518
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0884518 50 g
AAL0884509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0884518 Supplier Thermo Scientific Chemicals Supplier No. L0884518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1678-93-9 | C10H20 | 140.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1678-93-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00001530
InChI Key GGBJHURWWWLEQH-UHFFFAOYSA-N
Synonym n-butylcyclohexane, 1-cyclohexylbutane, cyclohexane, butyl, butyl-cyclohexane, butane, 1-cyclohexyl, n-butyl-cyclohexane, 1-cyclohexyl-butane, cyclohexane, n-butyl, acmc-20aj43, butylcyclohexane
PubChem CID 15506
IUPAC Name butylcyclohexane
SMILES CCCCC1CCCCC1

Specifications

Melting Point -78°C
Density 0.818
Boiling Point 178°C to 180°C
Flash Point 50°C (122°F)
Refractive Index 1.441
Quantity 50 g
UN Number UN3295
Beilstein 1900631
Formula Weight 140.27
Percent Purity 99%
Chemical Name or Material n-Butylcyclohexane
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 216-837-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only