Learn More
n-Butylamine, 99+%
CAS: 109-73-9 | C4H11N | 73.13 g/mol
$126.99 - $126.99
Chemical Identifiers
| CAS | 109-73-9 |
|---|---|
| Molecular Formula | C4H11N |
| Molecular Weight (g/mol) | 73.13 |
| MDL Number | MFCD00011690 |
| InChI Key | HQABUPZFAYXKJW-UHFFFAOYSA-N |
| Synonym | butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan |
| PubChem CID | 8007 |
| ChEBI | CHEBI:43799 |
| IUPAC Name | butan-1-amine |
| SMILES | CCCCN |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC107800010
|
Thermo Scientific Chemicals
107800010 |
1 L | Glass bottle |
Each for $126.99
Case of 4 Each for $0.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 109-73-9 | |
| 73.13 | |
| HQABUPZFAYXKJW-UHFFFAOYSA-N | |
| 8007 | |
| butan-1-amine |
| C4H11N | |
| MFCD00011690 | |
| butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan | |
| CHEBI:43799 | |
| CCCCN |
Specifications
| 109-73-9 | |
| 100.0 | |
| 0.7400g/mL | |
| −7°C | |
| 99% min. (GC) | |
| C4H11N | |
| CH3(CH2)3NH2 | |
| 1 L | |
| 04,53 | |
| 15,1545 | |
| Solubility in water: miscible with water. Other solubilities: miscible with alcohol and ether | |
| CCCCN | |
| 73.13 | |
| 0.5 mPa.s (20°C) | |
| 73.13 | |
| Liquid |
| 99.0 | |
| -49.0°C | |
| 78.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4000 to 1.4020 | |
| MFCD00011690 | |
| 04,156 | |
| 0.74 | |
| butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan | |
| HQABUPZFAYXKJW-UHFFFAOYSA-N | |
| butan-1-amine | |
| 8007 | |
| CHEBI:43799 | |
| 99+% | |
| n-Butylamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 203-699-2
RUO – Research Use Only