missing translation for 'onlineSavingsMsg'
Learn More

n-Butylamine, 99+%

Catalog No. AC107800010
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107800010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC107800010 Supplier Thermo Scientific Chemicals Supplier No. 107800010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-73-9 | C4H11N | 73.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-73-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.13
MDL Number MFCD00011690
InChI Key HQABUPZFAYXKJW-UHFFFAOYSA-N
Synonym butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan
PubChem CID 8007
ChEBI CHEBI:43799
IUPAC Name butan-1-amine
SMILES CCCCN

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -49.0°C
Density 0.7400g/mL
Boiling Point 78.0°C
Flash Point −7°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3NH2
Refractive Index 1.4000 to 1.4020
Quantity 1 L
Beilstein 04,156
Fieser 04,53
Specific Gravity 0.74
Merck Index 15,1545
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with alcohol and ether
Viscosity 0.5 mPa.s (20°C)
Formula Weight 73.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Butylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
EINECSNumber 203-699-2

RUO – Research Use Only