missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl nitrite, 95%

Catalog No. AAH5650136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5650136 500 g
AAH5650122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5650136 Supplier Thermo Scientific Chemicals Supplier No. H5650136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 544-16-1 | C4H9NO2 | 103.121 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 544-16-1
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002058
InChI Key JQJPBYFTQAANLE-UHFFFAOYSA-N
Synonym 1-butyl nitrite, nitrous acid, butyl ester, n-butyl nitrite, nitrous acid, n-butyl ester, nitrous acid butyl ester, n-butylnitrite, butylnitrite, unii-44p8qg0f3t, ccris 838, n-c4h9ono
PubChem CID 10996
IUPAC Name butyl nitrite
SMILES CCCCON=O

Specifications

Density 0.882
Boiling Point 78°C
Flash Point −13°C (9°F)
Refractive Index 1.376
Linear Formula CH3(CH2)3ONO
Quantity 500 g
UN Number UN2351
Beilstein 1701036
Solubility Information Slightly soluble in water.
Formula Weight 103.12
Percent Purity 95%
Chemical Name or Material n-Butyl nitrite
Show More Show Less
Hazard Category H225-H301+H331
Hazard Statement GHS H Statement
H225-H301-H331-H272
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic if inhaled.
May intensify fire; oxidizer.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL NITRITES
EINECSNumber 208-862-1
RTECSNumber RA0780000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only