Learn More
n-Butyl lactate, 99%
CAS: 138-22-7 | C7H14O3 | 146.19 g/mol
Supplier: Thermo Scientific Chemicals 250180025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| n-Butyl lactate | |
| 138-22-7 | |
| 100.0 | |
| Colorless | |
| 185.0°C to 187.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4200 to 1.4220 | |
| 2.5 L | |
| butyl lactate, n-butyl lactate, lactic acid, butyl ester, propanoic acid, 2-hydroxy-, butyl ester, butyllactate, 2-hydroxypropanoic acid butyl ester, butylester kyseliny mlecne, butyl alpha-hydroxypropionate, butyl lactate natural, fema no. 2205 | |
| MRABAEUHTLLEML-UHFFFAOYNA-N | |
| butyl 2-hydroxypropanoate | |
| 8738 | |
| 99% |
| 99% | |
| 98.5 | |
| -43.0°C | |
| 0.9800g/mL | |
| 71°C | |
| 99% | |
| C7H14O3 | |
| MFCD00004519 | |
| 0.98 | |
| Solubility in water: 42g/L (25 c). Other solubilities: hydrolyses in acids and alkalies,miscible with ether alcohol | |
| CCCCOC(=O)C(C)O | |
| 146.19 | |
| 146.19 | |
| Liquid |
Chemical Identifiers
| 138-22-7 | |
| 146.19 | |
| MRABAEUHTLLEML-UHFFFAOYNA-N | |
| 8738 | |
| CCCCOC(=O)C(C)O |
| C7H14O3 | |
| MFCD00004519 | |
| butyl lactate, n-butyl lactate, lactic acid, butyl ester, propanoic acid, 2-hydroxy-, butyl ester, butyllactate, 2-hydroxypropanoic acid butyl ester, butylester kyseliny mlecne, butyl alpha-hydroxypropionate, butyl lactate natural, fema no. 2205 | |
| butyl 2-hydroxypropanoate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 205-316-4
RTECSNumber : OD4025000
TSCA : TSCA
RUO – Research Use Only