missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl lactate, 99%

Catalog No. AC250185000
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC250185000 500 mL Glass bottle
AC250180025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC250185000 Supplier Thermo Scientific Chemicals Supplier No. 250185000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 138-22-7 | C7H14O3 | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 138-22-7
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.19
MDL Number MFCD00004519
InChI Key MRABAEUHTLLEML-UHFFFAOYNA-N
Synonym butyl lactate, n-butyl lactate, lactic acid, butyl ester, propanoic acid, 2-hydroxy-, butyl ester, butyllactate, 2-hydroxypropanoic acid butyl ester, butylester kyseliny mlecne, butyl alpha-hydroxypropionate, butyl lactate natural, fema no. 2205
PubChem CID 8738
IUPAC Name butyl 2-hydroxypropanoate
SMILES CCCCOC(=O)C(C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -43.0°C
Color Colorless
Density 0.9800g/mL
Boiling Point 185.0°C to 187.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4200 to 1.4220
Quantity 500 mL
Specific Gravity 0.98
Solubility Information Solubility in water: 42g/L (25 c). Other solubilities: hydrolyses in acids and alkalies,miscible with ether alcohol
Formula Weight 146.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Butyl lactate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 205-316-4
RTECSNumber OD4025000
TSCA TSCA

RUO – Research Use Only