missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl dodecanoate, 99+%

Catalog No. AA42024AP
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42024AP 500 mL
AA42024AC 25 mL
AA42024AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42024AP Supplier Thermo Scientific Chemicals Supplier No. 042024AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-18-3 | C16H32O2 | 256.43 g/mol

n-Butyl dodecanoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-18-3
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00042870
InChI Key NDKYEUQMPZIGFN-UHFFFAOYSA-N
Synonym butyl laurate, dodecanoic acid, butyl ester, n-butyl laurate, bytyl laurate, butyl dodecylate, lauric acid butyl ester, lauric acid, butyl ester, lauric acid n-butyl ester, unii-23985ocm4h, n-butyl n-dodecanoate
PubChem CID 61015
IUPAC Name butyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCCC

Specifications

Melting Point -10°C
Density 0.86 g/mL
Boiling Point 166°C
Flash Point >110°C (230°F)
Refractive Index 1.435
Quantity 500 mL
Beilstein 1776360
Solubility Information Soluble in water,0.06006mg/L @ 25°C.
Formula Weight 256.4
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material n-Butyl dodecanoate
Show More Show Less
EINECSNumber 203-370-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only