missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Chloride (HPLC), Fisher Chemical™

Catalog No. p-78514
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass
Safe-Cote™ Amber Glass 
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
B4294 4 L Amber Glass
B429SK4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4294 Supplier Thermo Fisher Scientific Supplier No. FLB4294

HPLC mobile phase for analytical and preparative separations

  • HPLC mobile phase for analytical and preparative separations.
  • High UV transmittance and low evaporation residue.
  • Filtered through a sub-micron filter to remove particulates.

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Absorbance 0.01 AU max., 0.01 AU max. at 254nm, 0.07 AU max. at 235nm, 0.33 AU max. at 225nm, 1.00 AU max. at 220nm
Melting Point -123°C
Color Colorless
Density 0.880g/cm³
Boiling Point 77°C
Flash Point −12°C
Assay Percent Range Assay: 99% min by GC
Packaging Amber Glass
Refractive Index 1.3985 to 1.4015 (at 25°C)
Linear Formula CH3(CH2)3Cl
Quantity 4 L
MDL Number MFCD00001009
UN Number 1127
Residue after Evaporation 1ppm max.
Vapor Pressure 108mbar at 20°C
Formula Weight 92.57
Percent Purity ≥99%
Grade Mobile phase for HPLC applications
Physical Form Liquid
Chemical Name or Material n-Butyl Chloride
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. The toxicological properties have not been fully investigated. Wear personal protective equipment. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. To avoid ignition of vapors by static electricity discharge, all metal parts of the equipment must b. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid