missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Chloride, B&J Brand™, for HPLC, GC and spectrophotometry, ≥99.5%, Honeywell Burdick & Jackson

Catalog No. p-7111728
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000097 6 x 1 L Glass Bottle
6000096 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000097 Supplier Honeywell Chemicals Supplier No. 0341L
Only null left

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

CAS Max % 100.0000%
Density 0.8862 g/cm3
Boiling Point 78°C
Flash Point −9°C
Loss on Drying 1 mg/L max.
Packaging Glass Bottle
Quantity 6 x 1 L
UN Number UN1127
Beilstein 1730909
Solubility Information 11% in Water at 20°C
Formula Weight 92.58
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material n-Butyl Chloride, B&J Brand™, for HPLC, GC and spectrophotometry, ≥99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Chlorobutanes