missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Alcohol, Purified (1-Butanol), Reagents

Catalog No. p-200021238
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337551 1 L
01337550 20 L
01337549 4 L
01337548 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337551 Supplier Labchem Inc Supplier No. C21200001C
Only null left

n-Butyl Alcohol, Purified (1-Butanol), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-Butanol, n-Butyl Alcohol, 1-Butyl Alcohol, Butan-1-ol
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point <88.6°C
Color Colorless
Boiling Point 118°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.81
Grade Pure
Chemical Name or Material n-Butyl Alcohol, Purified (1-Butanol)