missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acrylate, 98+%, stab. with up to 50ppm 4-methoxyphenol

Catalog No. p-7043919
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL03476AE 100 mL
AAL03476AP 500 mL
AAL034760F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL03476AE Supplier Thermo Scientific Chemicals Supplier No. L03476AE
Only null left

CAS: 141-32-2 | C7H12O2 | 128.171 g/mol

n-Butyl acrylate is used in polymer manufacturing, textile, leather finishing, formulating paints and adhesive. It is also used in the production of homopolymers and co-polymers like acrylic acid and its salts, esters, amides, methacrylates, acrylonitrile, maleates, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene and unsaturated polyesters. Further, it is also used in making vitamin B1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butyl acrylate is used in polymer manufacturing, textile, leather finishing, formulating paints and adhesive. It is also used in the production of homopolymers and co-polymers like acrylic acid and its salts, esters, amides, methacrylates, acrylonitrile, maleates, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene and unsaturated polyesters. Further, it is also used in making vitamin B1.

Solubility
Miscible with ethanol, ethyl ether and acetone. Slightly miscible with water and carbon tetrachloride.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, catalytic metals, organotin catalysts, amines, peroxides and halogens.

Chemical Identifiers

CAS 141-32-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00009446
InChI Key CQEYYJKEWSMYFG-UHFFFAOYSA-N
Synonym butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove
PubChem CID 8846
ChEBI CHEBI:3245
IUPAC Name butyl prop-2-enoate
SMILES CCCCOC(=O)C=C

Specifications

Melting Point -64°C
Density 0.895
Boiling Point 145°C to 146°C
Flash Point 39°C (102°F)
Odor Fruit-like
Refractive Index 1.418
Linear Formula CH2=CHCO2CH2CH2CH2CH3
Quantity 100 mL
UN Number UN2348
Beilstein 1749970
Sensitivity Light sensitive
Merck Index 14,1539
Solubility Information Miscible with ethanol,ethyl ether and acetone. Slightly miscible with water and carbon tetrachloride.
Formula Weight 128.17
Percent Purity ≥98%
Chemical Name or Material n-Butyl acrylate, stabilized with up to 50ppm 4-methoxyphenol
Show More Show Less
Hazard Category H226-H315-H317-H319-H332-H335
Hazard Statement GHS H Statement
H226-H315-H319-H317-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL ACRYLATES, STABILIZED
EINECSNumber 205-480-7
RTECSNumber UD3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only