missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Acetate (Reagent), Fisher Chemical™

Catalog No. p-78486
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
B3964 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3964 Supplier Thermo Fisher Scientific Supplier No. BPB3964
Only null left

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Boiling Range 5°C max.
Melting Point -77.9°C
Color Colorless
Density 0.880g/cm³
pH 6.2
Boiling Point 125°C
Flash Point 22°C
Packaging Glass Bottle
Linear Formula CH3COOCH2(CH2)2CH3
Quantity 4 L
UN Number 1123
Vapor Pressure 13mbar at 20°C
Formula Weight 116.16
Grade Reagent
Physical Form Liquid
Chemical Name or Material n-Butyl Acetate
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause pulmonary edema. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid