missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyl Acetate ASTM D4615, Reagents
n-Butyl Acetate ASTM D4615, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C21563004C
Specifications
| n-Butyl Acetate ASTM D4615 | |
| 126°C | |
| Colorless | |
| -78°C | |
| C6H12O2 | |
| 0.88 | |
| DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
| butyl acetate | |
| Liquid |
| 123-86-4 | |
| -78°C | |
| 126°C | |
| Glass Bottle | |
| 4 L | |
| Butyl Ethanoate, Acetic Acid Butyl Ester | |
| CCCCOC(C)=O | |
| 116.16 |
Chemical Identifiers
| 123-86-4 | |
| 116.16 | |
| Butyl Ethanoate, Acetic Acid Butyl Ester | |
| CCCCOC(C)=O |
| C6H12O2 | |
| DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
| butyl acetate |