missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Acetate ASTM D4615, Reagents

Catalog No. 01337381
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01337381 4 L Poly Bottle
01337380 4 L Glass Bottle
01337379 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337381 Supplier Labchem Inc Supplier No. C21563004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

n-Butyl Acetate ASTM D4615, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym Butyl Ethanoate, Acetic Acid Butyl Ester
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Boiling Range 126°C
Melting Point -78°C
Color Colorless
Boiling Point 126°C
Melting Range -78°C
Packaging Poly Bottle
Quantity 4 L
Specific Gravity 0.88
Physical Form Liquid
Chemical Name or Material n-Butyl Acetate ASTM D4615