missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Acetate, ACS, 99.5%, Spectrum™ Chemical

Catalog No. 18611911
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
Steel Can
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611911 500 mL Amber Glass Bottle
18611908 100 mL Amber Glass Bottle
18611905 1 L Amber Glass Bottle
18611909 2.5 L Poly-Coated Glass Bottle
18611906 4 L Amber Glass Bottle
18611907 20 L Steel Can
18611910 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18611911 Supplier Spectrum Chemical Mfg Cor Supplier No. B1182500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Residue after Evaporation 1e-05
Percent Purity 0.995
Grade ACS
Titratable Acid 0.0016 meq/g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.