missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Acetate, ACS, 99.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B118225LTPV

 View more versions of this product

Catalog No. 18611909

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

123-86-4
Poly-Coated Glass Bottle
2.5 L
DKPFZGUDAPQIHT-UHFFFAOYSA-N
butyl acetate
0.995
0.0016 meq/g
1
C6H12O2
1e-05
CCCCOC(C)=O
116.16
ACS

Chemical Identifiers

123-86-4
116.16
butyl acetate
C6H12O2
DKPFZGUDAPQIHT-UHFFFAOYSA-N
CCCCOC(C)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.