missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl acetate, 99+%, for spectroscopy

Catalog No. AC167685000
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167685000 500 mL Glass bottle
AC167680010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167685000 Supplier Thermo Scientific Chemicals Supplier No. 167685000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Absorbance (1 cm cell vs water), 0.01 max. at 320nm, 0.02 max. at 300nm, 0.04 max. at 275nm, 0.15 max. at 260nm, 0.4 max. at 256nm, 1 max. at 254nm
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3CO2(CH2)3CH3
Quantity 500 mL
Residue after Evaporation 0.0005% max.
Beilstein 02, 130
Merck Index 15, 1537
Vapor Pressure 13mbar at 20°C
Formula Weight 116.16
Percent Purity ≥99.0%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material n-Butyl acetate, 99+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-658-1
RTECSNumber AF7350000
TSCA TSCA

RUO – Research Use Only