missing translation for 'onlineSavingsMsg'
Learn More

N-Bsmoc-glycine, 99%

Catalog No. p-7047774
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1973703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1973703 Supplier Thermo Scientific Chemicals Supplier No. L1973703

CAS: 197245-13-9 | C12H11NO6S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 197245-13-9
Molecular Formula C12H11NO6S
MDL Number MFCD03792135
Synonym n-bsmoc-glycine, n-benzo b thiophenesulfone-2-methoxycarbonyl glycine, n-benzo b thiophenesulfone-2-methoxycarbonyl aminoacetic acid, 1,1-dioxo-1??-benzothiophen-3-yl methoxy carbonyl amino acetic acid

Specifications

Melting Point 161°C to 163°C
Beilstein 8269937
Quantity 1 g
Formula Weight 297.29
Percent Purity 99%
Chemical Name or Material N-Bsmoc-glycine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only