missing translation for 'onlineSavingsMsg'
Learn More

N-Bromosuccinimide, 98%, Spectrum™ Chemical

Catalog No. p-7157891
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604760 100 g Amber Glass Bottle
18604761 500 g Amber Glass Bottle
18604762 2.5 kg Poly Pail
18604763 25 kg Fiber Drum
18604764 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604760 Supplier Spectrum Chemical Mfg Cor Supplier No. B1158100GM
Only null left

B1158, 128-08-5, C4H4BrNO2

Spectrum™ Chemical N-Bromosuccinimide , also known as NBS, is a chemical reagent used in organic chemistry in electrophilic addition reactions and radical substitution. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 128-08-5
Molecular Formula C4H4BrNO2
Molecular Weight (g/mol) 177.99
InChI Key PCLIMKBDDGJMGD-UHFFFAOYSA-N
IUPAC Name 1-bromopyrrolidine-2,5-dione
SMILES BrN1C(=O)CCC1=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98%
Grade Reagent