missing translation for 'onlineSavingsMsg'
Learn More

N-Bromosuccinimide, 98%, Spectrum™ Chemical

Catalog No. 18604764
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604764 50 kg Amber Glass Bottle
18604760 100 g Amber Glass Bottle
18604761 500 g Amber Glass Bottle
18604762 2.5 kg Amber Glass Bottle
18604763 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604764 Supplier Spectrum Chemical Mfg Cor Supplier No. B115850KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

B1158, 128-08-5, C4H4BrNO2

Spectrum™ Chemical N-Bromosuccinimide , also known as NBS, is a chemical reagent used in organic chemistry in electrophilic addition reactions and radical substitution. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 128-08-5
Molecular Formula C4H4BrNO2
Molecular Weight (g/mol) 177.99
InChI Key PCLIMKBDDGJMGD-UHFFFAOYSA-N
IUPAC Name 1-bromopyrrolidine-2,5-dione
SMILES BrN1C(=O)CCC1=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 50 kg
Percent Purity 98%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material N-Bromosuccinimide