missing translation for 'onlineSavingsMsg'
Learn More

N-Bromosuccinimide, 98%, Spectrum™ Chemical

Catalog No. 18604760
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604760 100 g Amber Glass Bottle
18604761 500 g Amber Glass Bottle
18604762 2.5 kg Amber Glass Bottle
18604763 25 kg Amber Glass Bottle
18604764 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604760 Supplier Spectrum Chemical Mfg Cor Supplier No. B1158100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

B1158, 128-08-5, C4H4BrNO2

Spectrum™ Chemical N-Bromosuccinimide , also known as NBS, is a chemical reagent used in organic chemistry in electrophilic addition reactions and radical substitution. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 128-08-5
Molecular Formula C4H4BrNO2
Molecular Weight (g/mol) 177.99
InChI Key PCLIMKBDDGJMGD-UHFFFAOYSA-N
IUPAC Name 1-bromopyrrolidine-2,5-dione
SMILES BrN1C(=O)CCC1=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material N-Bromosuccinimide