missing translation for 'onlineSavingsMsg'
Learn More

N-Bromosaccharin 98.0+%, TCI America™

Catalog No. B21525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21525G 5 g
B215225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21525G Supplier TCI America Supplier No. B21525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35812-01-2
Molecular Formula C7H4BrNO3S
Molecular Weight (g/mol) 262.08
MDL Number MFCD03844772
InChI Key QRADPXNAURXMSB-UHFFFAOYSA-N
Synonym n-bromosaccharin, 2-bromobenzo d isothiazol-3 2h-one 1,1-dioxide, 2-bromo-1,2-benzisothiazol-3 2h-one 1,1-dioxide, 2-bromo-1??,2-benzothiazole-1,1,3-trione
PubChem CID 64791
IUPAC Name 2-bromo-2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
SMILES BrN1C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 262.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Bromosaccharin
TSCA No
Recommended Storage Refrigerator