missing translation for 'onlineSavingsMsg'
Learn More

N-Bromophthalimide 95.0+%, TCI America™

Catalog No. p-7119209
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16975G 5 g
B169725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16975G Supplier TCI America Supplier No. B16975G
Only null left

Chemical Identifiers

CAS 2439-85-2
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00005888
InChI Key MARXMDRWROUXMD-UHFFFAOYSA-N
PubChem CID 75542
IUPAC Name 2-bromoisoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)Br

Specifications

Melting Point 207°C
Quantity 5 g
Synonym n-bromophthalimide, 2-bromoisoindoline-1,3-dione, 2-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-bromo, n-bromophtalimide, n-bromo phthalimide, acmc-1cjlt, 2-bromobenzo c azoline-1,3-dione, 1h-isoindole-1,3 2h-dione,2-bromo
Formula Weight 226.03
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Bromophthalimide
EINECSNumber (5)-0080
TSCA No