missing translation for 'onlineSavingsMsg'
Learn More

N-(Bromomethyl)phthalimide 96.0+%, TCI America™

Catalog No. p-7134997
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11925G 5 g
P119225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11925G Supplier TCI America Supplier No. P11925G
Only null left

Chemical Identifiers

CAS 5332-26-3
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
MDL Number MFCD00005897
InChI Key UUSLLECLCKTJQF-UHFFFAOYSA-N
Synonym n-bromomethyl phthalimide, 2-bromomethyl isoindoline-1,3-dione, n-bromomethylphthalimide, phthalimide, n-bromomethyl, phthalimidomethyl bromide, 1h-isoindole-1,3 2h-dione, 2-bromomethyl, 2-bromomethyl isoindole-1,3-dione, 2-bromomethyl-1h-isoindole-1,3 2h-dione, 2-bromomethyl-2,3-dihydro-1h-isoindole-1,3-dione, acmc-1awzk
PubChem CID 79244
IUPAC Name 2-(bromomethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CBr

Specifications

Melting Point 151°C
Quantity 5 g
Formula Weight 240.06
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(Bromomethyl)phthalimide