missing translation for 'onlineSavingsMsg'
Learn More

N-(Bromomethyl)phthalimide, 95%

Catalog No. p-7043805
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0281809 10 g
AAL0281818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0281809 Supplier Thermo Scientific Chemicals Supplier No. L0281809
Only null left

CAS: 5332-26-3 | C9H6BrNO2 | 240.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5332-26-3
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
MDL Number MFCD00005897
InChI Key UUSLLECLCKTJQF-UHFFFAOYSA-N
Synonym n-bromomethyl phthalimide, 2-bromomethyl isoindoline-1,3-dione, n-bromomethylphthalimide, phthalimide, n-bromomethyl, phthalimidomethyl bromide, 1h-isoindole-1,3 2h-dione, 2-bromomethyl, 2-bromomethyl isoindole-1,3-dione, 2-bromomethyl-1h-isoindole-1,3 2h-dione, 2-bromomethyl-2,3-dihydro-1h-isoindole-1,3-dione, acmc-1awzk
PubChem CID 79244
IUPAC Name 2-(bromomethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CBr

Specifications

Melting Point 144°C to 150°C
Odor Odorless
Quantity 10 g
Beilstein 140943
Sensitivity Light and moisture sensitive
Formula Weight 240.07
Percent Purity 95%
Chemical Name or Material N-(Bromomethyl)phthalimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-239-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only