missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-trans-4-Hydroxy-L-proline methyl ester, 97%, Thermo Scientific Chemicals

$232.38 - $232.38

Chemical Identifiers

CAS 74844-91-0
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.28
MDL Number MFCD00076981
InChI Key MZMNEDXVUJLQAF-SFYZADRCSA-N
Synonym boc-hyp-ome, n-boc-trans-4-hydroxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, boc-l-hydroxyproline methyl ester, boc-hydroxyproline-ome, methyl trans-1-boc-4-hydroxy-l-prolinate, n-tert-butoxycarbonyl-trans-4-hydroxy-l-proline methyl ester, trans-1-tert-butoxycarbonyl-4-hydroxy-l-proline methyl ester, n-boc-trans-4-hydroxyl-l-proline methyl ester
PubChem CID 2734883
SMILES COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC376250050
SDS View Documents
Thermo Scientific Chemicals
376250050
5 g Glass bottle
Each for $232.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

74844-91-0
245.28
MZMNEDXVUJLQAF-SFYZADRCSA-N
2734883
C11H19NO5
MFCD00076981
boc-hyp-ome, n-boc-trans-4-hydroxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, boc-l-hydroxyproline methyl ester, boc-hydroxyproline-ome, methyl trans-1-boc-4-hydroxy-l-prolinate, n-tert-butoxycarbonyl-trans-4-hydroxy-l-proline methyl ester, trans-1-tert-butoxycarbonyl-4-hydroxy-l-proline methyl ester, n-boc-trans-4-hydroxyl-l-proline methyl ester
COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C

Specifications

74844-91-0
100.0
White to Beige
96% min. (GC)
C11H19NO5
5 g
− 65.00
Solubility in water: slightly soluble.
COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C
245.28
245.28
Crystalline Powder
96.0
91.0°C to 96.0°C
Authentic
Glass bottle
MFCD00076981
−65° (20°C c=1 Chloroform)
boc-hyp-ome, n-boc-trans-4-hydroxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, boc-l-hydroxyproline methyl ester, boc-hydroxyproline-ome, methyl trans-1-boc-4-hydroxy-l-prolinate, n-tert-butoxycarbonyl-trans-4-hydroxy-l-proline methyl ester, trans-1-tert-butoxycarbonyl-4-hydroxy-l-proline methyl ester, n-boc-trans-4-hydroxyl-l-proline methyl ester
MZMNEDXVUJLQAF-SFYZADRCSA-N
1-O-tert-butyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
2734883
97%
N-BOC-trans-4-Hydroxy-L-proline methyl ester
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only