missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-trans-4-hydroxy-L-proline, 97%

CAS: 13726-69-7 | C10H17NO5 | 231.25 g/mol

$166.66 - $496.05

Chemical Identifiers

CAS 13726-69-7
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.25
MDL Number MFCD00053370
InChI Key BENKAPCDIOILGV-UHFFFAOYNA-N
Synonym boc-hyp-oh, boc-l-hydroxyproline, 2s,4r-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, boc-trans-4-hydroxy-l-proline, n-boc-trans-4-hydroxy-l-proline, boc-l-hyp-oh, boc-hydroxyproline, unii-rc99p5b8ig, trans-boc-hyp-oh, 4r-1-tert-butoxycarbonyl-4-hydroxy-l-proline
PubChem CID 88804
IUPAC Name (2S,4R)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2711006
View Documents
Thermo Scientific Chemicals
H2711006
5 g
Each for $166.66
Only null left
 
AAH2711014
View Documents
Thermo Scientific Chemicals
H2711014
25 g
Each for $496.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13726-69-7
231.25
BENKAPCDIOILGV-UHFFFAOYNA-N
88804
CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O
C10H17NO5
MFCD00053370
boc-hyp-oh, boc-l-hydroxyproline, 2s,4r-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, boc-trans-4-hydroxy-l-proline, n-boc-trans-4-hydroxy-l-proline, boc-l-hyp-oh, boc-hydroxyproline, unii-rc99p5b8ig, trans-boc-hyp-oh, 4r-1-tert-butoxycarbonyl-4-hydroxy-l-proline
(2S,4R)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid

Specifications

13726-69-7
C10H17NO5
5 g
BENKAPCDIOILGV-UHFFFAOYNA-N
(2S,4R)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
88804
97%
123°C to 127°C
MFCD00053370
boc-hyp-oh, boc-l-hydroxyproline, 2s,4r-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, boc-trans-4-hydroxy-l-proline, n-boc-trans-4-hydroxy-l-proline, boc-l-hyp-oh, boc-hydroxyproline, unii-rc99p5b8ig, trans-boc-hyp-oh, 4r-1-tert-butoxycarbonyl-4-hydroxy-l-proline
CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O
231.25
231.25
N-Boc-trans-4-hydroxy-L-proline
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only