missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-trans-4-hydroxy-D-proline, 99%, Thermo Scientific Chemicals

Catalog No. AAH2715503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2715503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2715503 Supplier Thermo Scientific Chemicals Supplier No. H2715503
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 147266-92-0
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.25
MDL Number MFCD01861341
InChI Key BENKAPCDIOILGV-UHFFFAOYNA-N
Synonym 2r,4s-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, n-boc-trans-4-hydroxy-d-proline, boc-trans-4-hydroxy-d-proline, boc-d-hyp-oh, unii-7k1v4sdl4l, 7k1v4sdl4l, 2r,4s-1-tert-butoxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 4s-1-tert-butoxycarbonyl-4-hydroxy-d-proline, n-boc-4-hydroxy-d-proline, trans, 2r,4s-n-alpha-t-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid
PubChem CID 688615
IUPAC Name (2R,4S)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O

Specifications

Melting Point ∼123°C
Quantity 1 g
Formula Weight 231.25
Percent Purity 99%
Chemical Name or Material N-Boc-trans-4-hydroxy-D-proline
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only