missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-sarcosine methyl ester, 97%, Thermo Scientific Chemicals

Catalog No. p-7054248
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6230406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6230406 Supplier Thermo Scientific Chemicals Supplier No. H6230406
Only null left

N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.

Notes
Incompatible with oxidizing agents. Heat sensitive.

Chemical Identifiers

CAS 42492-57-9
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD04973115
InChI Key LTBDTYJYKRLTRN-UHFFFAOYSA-N
Synonym methyl 2-tert-butoxycarbonyl methyl amino acetate, n-boc-sarcosine methyl ester, boc-gly-ome, boc-sarcosine methyl ester, ltbdtyjykrltrn-uhfffaoysa, n-t-butoxycarbonyl-sarcosine methyl ester, n-tert-butoxycarbonylsarcosine methyl ester, methyl n-tert-butoxycarbonyl-n-methylglycinate, methyl 2-tert-butoxy carbonyl methyl amino acetate
PubChem CID 14018930
SMILES COC(=O)CN(C)C(=O)OC(C)(C)C

Specifications

Color Yellow
Linear Formula (CH3)3COCON(CH3)CH2CO2CH3
IUPAC Name methyl 2-{[(tert-butoxy)carbonyl](methyl)amino}acetate
Quantity 5 g
Formula Weight 203.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Boc-sarcosine methyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only