missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-S-benzyl-L-cysteine, 98%

Catalog No. AAH6373614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6373614 25 g
AAH6373606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6373614 Supplier Thermo Scientific Chemicals Supplier No. H6373614
Only null left

CAS: 5068-28-0 | C15H21NO4S | 311.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5068-28-0
Molecular Formula C15H21NO4S
Molecular Weight (g/mol) 311.40
MDL Number MFCD00065567
InChI Key IFVORPLRHYROAA-UHFFFAOYNA-N
Synonym boc-s-benzyl-l-cysteine, boc-cys bzl-oh, r-3-benzylthio-2-tert-butoxycarbonyl amino propanoic acid, n-boc-s-benzyl-l-cysteine, n-tert-butoxycarbonyl-s-benzyl-l-cysteine, s-benzyl-n-tert-butoxycarbonyl-l-cysteine, 2r-3-benzylsulfanyl-2-tert-butoxycarbonyl amino propanoic acid, boc-s-benzyl-d-cys, boc-s-benzyl-l-cys, alanine, 3-benzylthio-n-carboxy-, n-tert-butyl ester, l
PubChem CID 2724757
SMILES CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 86°C to 88°C
Beilstein 2000417
IUPAC Name (2R)-3-benzylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Quantity 25 g
Formula Weight 311.4
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-S-benzyl-L-cysteine
EINECSNumber 225-772-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only