missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Pyrrolidine, 97%

Catalog No. AC425820050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425820050 5 g Glass bottle
AC425820250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425820050 Supplier Thermo Scientific Chemicals Supplier No. 425820050

CAS: 86953-79-9 | C9H17NO2 | 171.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86953-79-9
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00216581
InChI Key LPQZERIRKRYGGM-UHFFFAOYSA-N
Synonym 1-boc-pyrrolidine, n-boc-pyrrolidine, pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 1-pyrrolidinecarboxylate, 1-tert-butoxycarbonyl pyrrolidine, 1-pyrrolidinecarboxylic acid tert-butyl ester, boc-pyrrolidine, acmc-20aivt, pubchem9422, 1-boc-3-hydropyrrolidine
PubChem CID 643455
IUPAC Name tert-butyl pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1

Specifications

Density 0.9770g/mL
Boiling Point 80°C (0.2 mmHg)
Flash Point 86°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.977
Formula Weight 171.24
Percent Purity 97%
Chemical Name or Material N-BOC-Pyrrolidine
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only