missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-O-tert-butyl-L-tyrosine, 98%
CAS: 47375-34-8 | C18H27NO5 | 337.42 g/mol
$264.79 - $921.49
Chemical Identifiers
| CAS | 47375-34-8 |
|---|---|
| Molecular Formula | C18H27NO5 |
| Molecular Weight (g/mol) | 337.42 |
| MDL Number | MFCD00065598 |
| InChI Key | ZEQLLMOXFVKKCN-YQTOOIBONA-N |
| Synonym | boc-tyr tbu-oh, boc-o-tert-butyl-l-tyrosine, boc-tyr tbul-oh, s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-tert-butyl-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-4-tert-butoxy phenyl propanoic acid, boc-tyr but-oh, pubchem12288 |
| PubChem CID | 7017901 |
| IUPAC Name | (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid |
| SMILES | CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6280106
|
Thermo Scientific Chemicals
H6280106 |
5 g |
Each for $264.79
|
|
|||||
|
AAH6280114
|
Thermo Scientific Chemicals
H6280114 |
25 g |
Each for $921.49
|
|
|||||
Description
N-Boc-O-tert-butyl-L-tyrosine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-O-tert-butyl-L-tyrosine is used as pharmaceutical intermediate.
Solubility
Soluble in DMF. Insoluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Chemical Identifiers
| 47375-34-8 | |
| 337.42 | |
| ZEQLLMOXFVKKCN-YQTOOIBONA-N | |
| 7017901 | |
| CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O |
| C18H27NO5 | |
| MFCD00065598 | |
| boc-tyr tbu-oh, boc-o-tert-butyl-l-tyrosine, boc-tyr tbul-oh, s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-tert-butyl-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-4-tert-butoxy phenyl propanoic acid, boc-tyr but-oh, pubchem12288 | |
| (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid |
Specifications
| 47375-34-8 | |
| C18H27NO5 | |
| 5 g | |
| boc-tyr tbu-oh, boc-o-tert-butyl-l-tyrosine, boc-tyr tbul-oh, s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-tert-butyl-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-4-tert-butoxy phenyl propanoic acid, boc-tyr but-oh, pubchem12288 | |
| ZEQLLMOXFVKKCN-YQTOOIBONA-N | |
| CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O | |
| 337.42 | |
| 337.41 | |
| N-Boc-O-tert-butyl-L-tyrosine |
| 113°C to 118°C | |
| MFCD00065598 | |
| 4327336 | |
| Soluble in DMF. Insoluble in water. | |
| −15° (c=1 in DMF) | |
| (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid | |
| 7017901 | |
| 98% |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only