missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-tert-butyl-L-threonine, 95%

CAS: 13734-40-2 | C13H25NO5 | 275.35 g/mol

$75.55 - $335.41

Chemical Identifiers

CAS 13734-40-2
Molecular Formula C13H25NO5
Molecular Weight (g/mol) 275.35
MDL Number MFCD00151115
InChI Key LKRXXARJBFBMCE-BDAKNGLRSA-N
Synonym boc-thr tbu-oh, boc-o-tert-butyl-l-threonine, boc-o-t-butyl-l-threonine, 2s,3r-3-tert-butoxy-2-tert-butoxycarbonyl amino butanoic acid, l-threonine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, boc-thr t-bu-oh, pubchem12289, boc-l-thr tbu-oh, boc-o-tert-butyl-l-thr-oh
PubChem CID 7015770
IUPAC Name (2S,3R)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6271303
View Documents
Thermo Scientific Chemicals
H6271303
1 g
Each for $75.55
Only null left
 
AAH6271306
View Documents
Thermo Scientific Chemicals
H6271306
5 g
Each for $335.41
Only null left
 
Description

Description

N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.

Solubility
Soluble in DMF.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

13734-40-2
275.35
LKRXXARJBFBMCE-BDAKNGLRSA-N
7015770
C[C@@H](OC(C)(C)C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
C13H25NO5
MFCD00151115
boc-thr tbu-oh, boc-o-tert-butyl-l-threonine, boc-o-t-butyl-l-threonine, 2s,3r-3-tert-butoxy-2-tert-butoxycarbonyl amino butanoic acid, l-threonine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, boc-thr t-bu-oh, pubchem12289, boc-l-thr tbu-oh, boc-o-tert-butyl-l-thr-oh
(2S,3R)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid

Specifications

95°C to 98°C
C13H25NO5
4454820
Soluble in DMF.
C[C@@H](OC(C)(C)C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
1 g
7015770
95%
N-Boc-O-tert-butyl-L-threonine
13734-40-2
MFCD00151115
boc-thr tbu-oh, boc-o-tert-butyl-l-threonine, boc-o-t-butyl-l-threonine, 2s,3r-3-tert-butoxy-2-tert-butoxycarbonyl amino butanoic acid, l-threonine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, boc-thr t-bu-oh, pubchem12289, boc-l-thr tbu-oh, boc-o-tert-butyl-l-thr-oh
LKRXXARJBFBMCE-BDAKNGLRSA-N
(2S,3R)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
275.35
275.34
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only