missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-tert-butyl-L-threonine, 95%

Catalog No. aah6271306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6271306 5 g
AAH6271303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6271306 Supplier Thermo Scientific Chemicals Supplier No. H6271306
Only null left

CAS: 13734-40-2 | C13H25NO5 | 275.35 g/mol

N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.

Solubility
Soluble in DMF.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 13734-40-2
Molecular Formula C13H25NO5
Molecular Weight (g/mol) 275.35
MDL Number MFCD00151115
InChI Key LKRXXARJBFBMCE-BDAKNGLRSA-N
Synonym boc-thr tbu-oh, boc-o-tert-butyl-l-threonine, boc-o-t-butyl-l-threonine, 2s,3r-3-tert-butoxy-2-tert-butoxycarbonyl amino butanoic acid, l-threonine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, boc-thr t-bu-oh, pubchem12289, boc-l-thr tbu-oh, boc-o-tert-butyl-l-thr-oh
PubChem CID 7015770
IUPAC Name (2S,3R)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 95°C to 98°C
Beilstein 4454820
Solubility Information Soluble in DMF.
Quantity 5 g
Formula Weight 275.34
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-O-tert-butyl-L-threonine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only