missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt, 98%

Catalog No. p-7053138
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5916306 5 g
AAH5916314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5916306 Supplier Thermo Scientific Chemicals Supplier No. H5916306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18942-50-2 | C24H46N2O5 | 442.641 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18942-50-2
Molecular Formula C24H46N2O5
Molecular Weight (g/mol) 442.641
MDL Number MFCD00065590
InChI Key AIEUUHIXSUNTGV-QRPNPIFTSA-N
Synonym boc-ser tbu-oh dcha, boc-ser tbu-oh.dcha, boc-o-tert-butyl-l-serine dicyclohexylamine salt, pubchem12161, boc-o-t-butyl-l-serine dcha salt, boc-o-tert-butyl-l-serine dcha salt, boc-ser tbu-oh dicyclohexylammonium salt, boc-o-tert-butyl-l-serine dicyclohexyl amine salt, boc-o-tert-butyl-l-serine dicyclohexylammonium salt, boc-ser tbu-oh dicyclohexylammonium salt n
PubChem CID 12786117
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)NC(=O)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2

Specifications

Melting Point 161°C to 164°C
Linear Formula (CH3)3COCH2CH(COOH)NHCOOC(CH3)3·C12H23N
Beilstein 5207271
Quantity 5 g
Formula Weight 442.63
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only