missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-methyl-L-tyrosine, 98%

Catalog No. AAH6182006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6182006 5 g
AAH6182003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6182006 Supplier Thermo Scientific Chemicals Supplier No. H6182006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 53267-93-9 | C15H21NO5 | 295.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53267-93-9
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.34
MDL Number MFCD00065603
InChI Key SLWWWZWJISHVOU-STGVRZAANA-N
Synonym boc-tyr me-oh, boc-o-methyl-l-tyrosine, 2s-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl propanoic acid, boc-4-methoxyphenylalanine, s-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl propanoic acid, boc-l-4-methoxyphenylalanine, boc-4-methoxy-phe-oh, boc-4-methoxy-l-phenylalanine, n-tert-butoxycarbonyl-o-methyltyrosine, n-alpha-t-butyloxycarbonyl-o-methyl-l-tyrosine
PubChem CID 2762280
IUPAC Name (2S)-3-(4-methoxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES COC1=CC=C(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)C=C1

Specifications

Quantity 5 g
Formula Weight 295.33
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-O-methyl-L-tyrosine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only